missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NRG1 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 2C6D12, Proteintech
Mouse Monoclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | NRG1 |
---|---|
Clone | 2C6D12 |
Concentration | 1 mg/mL |
Applications | Immunofluorescence, Immunocytochemistry, Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16888415
|
Proteintech
66492-1-IG-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16878415
|
Proteintech
66492-1-IG-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The neuregulins are a family of ErbB/HER ligands encoded by four genes. Neuregulin-1 gene, NRG1, encodes numerous splice variants with differing transcription initiation sites. Neuregulin-1 includes a range of isoforms with varying glycosylation, regulation of expression and function. Neuregulin-1 splice variants each bear an EGF-like domain, though otherwise have unique domain structures, differing functions and discrete tissue distribution. Six types of Neuregulin-1 isoform groups have been defined based on their structural features. Three types are most often described, type I (ARIA, NDF or HRG), type II (GGF) and type III (SMDF). Neuregulin-1 has been linked to schizophrenia and has diverse neural functions. Neuregulin-1 affects cell migration, the differentiation of neural crest and Schwann cells, and acts to upregulate the expression of acetylcholine receptors at muscle fibers during the formation of neuromuscular junctions.Specifications
NRG1 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
ARIA, GGF, GGF2, Glial growth factor, Heregulin, HGL, HRG, HRG1, HRGA, NDF, Neu differentiation factor, neuregulin 1, NRG1, Pro NRG1, SMDF | |
NRG1 | |
IgG1 | |
Protein G | |
Antibody |
2C6D12 | |
Immunofluorescence, Immunocytochemistry, Western Blot | |
Unconjugated | |
Mouse | |
Human, Rat, Pig, Mouse | |
P43322, Q02297 | |
102164569, 112400, 211323, 3084 | |
NRG1 Fusion Protein Ag0803 | |
Primary | |
-20°C | |
NRG1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
NRG1 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 2C6D12, Proteintech