missing translation for 'onlineSavingsMsg'
Learn More

NT5DC3 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 26242-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16884755

  • 1355.00 SEK / 20µL
Estimated Shipment: 22-03-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

NT5DC3 gene ontology annotations related to this gene include 5′-nucleotidase activity; metal ion binding.
Specifications

Specifications

NT5DC3
Polyclonal
Unconjugated
NT5DC3
GNN, GRP94 neighboring nucleotidase, NT5DC3, TU12B1 TY, TU12B1TY
Rabbit
Antigen Affinity Chromatography
RUO
51559
-20°C
Liquid
Immunoprecipitation
0.5 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q86UY8
NT5DC3
NT5DC3 Fusion Protein Ag23962
20 μL
Primary
Human
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers