Learn More
Thermo Scientific™ Nunc™ MicroWell™ 96-Well, Nunclon Delta-Treated, Flat-Bottom Microplate
Color: Clear; Sterility: Sterile; Lid: Without Lid
1560.00 SEK - 4950.00 SEK
Specifications
Barcode | Custom Barcoding Available |
---|---|
Certifications/Compliance | All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) |
Lid | Without Lid |
Array | 8 x 12 |
Product Line | Nunc |
Product Code | Brand | No. of Wells | No. per Pack | No. per Case | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Wells | No. per Pack | No. per Case | Price | Quantity & Availability | ||||
10058820
![]() |
Thermo Scientific™
168055 |
96 | 1 Microplate | 50 Microplates |
1560.00 SEK
Pack of 50 |
In Stock
Log in to see stock. |
||||
10214171
![]() |
Thermo Scientific™
156545 |
96 | 10 Microplates | 180 Microplates |
4950.00 SEK
Pack of 180 |
In Stock
Log in to see stock. |
||||
Description
Thermo Scientific™ Nunc MicroWell 96-Well Microplates are optimized for robotics, automated readers and liquid handling systems. 96-well microplates are used for antibiotic screens, cell-based assays and screening compounds. The flat bottom shape is ideal for microscopic and optical measurements. Our plates are high quality for consistent and reliable results.
- Clear polystyrene has excellent optical properties making it ideal for optical measurements
- Untreated polystyrene is hydrophobic and is suited for growth of suspension cultures that can proliferate and grow without attachment
- Untreated surfaces are ideal for adding coatings in your lab
- Cell Culture Treated: The Nunclon™ Delta cell culture treated surface is a hydrophilic surface that promotes cell attachment and growth
- Plates are optimized for robotics, plate readers and liquid handlers
- Customized bar coding is available
Specifications
Custom Barcoding Available | |
Without Lid | |
Nunc | |
Microplate | |
Polystyrene | |
96 | |
Nunclon Delta |
All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) | |
8 x 12 | |
Clear | |
High Flange | |
400 ÎĽL | |
Sterile | |
Flat Bottom (F-well) |