Learn More
NUP50 Mouse anti-Human, Unconjugated, Clone: 1D5A7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67001-1-IG-150UL
Description
The NUP50 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NUP50. This antibody recognizes human antigen. The NUP50 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
NUP50 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, Immunohistochemistry, ELISA | |
1D5A7 | |
Unconjugated | |
150 μL | |
10762 | |
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.