missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUP50 Mouse anti-Human, Unconjugated, Clone: 1D5A7, Proteintech
Mouse Monoclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | NUP50 |
---|---|
Clone | 1D5A7 |
Concentration | 1000 ÎĽg/mL |
Applications | Western Blot, Immunohistochemistry, ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16889355
|
Proteintech
67001-1-IG-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16879355
|
Proteintech
67001-1-IG-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The NUP50 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NUP50. This antibody recognizes human antigen. The NUP50 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
NUP50 | |
1000 ÎĽg/mL | |
Monoclonal | |
Mouse | |
10762 | |
Primary |
1D5A7 | |
Western Blot, Immunohistochemistry, ELISA | |
Unconjugated | |
Human | |
IgG1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
NUP50 Mouse anti-Human, Unconjugated, Clone: 1D5A7, Proteintech