missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NXF1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10328-1-AP-20UL
This item is not returnable.
View return policy
Description
p115 (Vesicle docking protein p115) is a peripheral membrane protein that is located on the Golgi complex. p115 exists as a homodimer with two globular heads, an extended coiled-coil tail, followed by an acidic domain at the extreme C terminus. p115 is homologous to a yeast protein, Uso1p, which is required for ER to Golgi transport. p115 likely plays an important role in vesicle transportation from the ER to the cis-Golgi compartments.Specifications
NXF1 | |
Polyclonal | |
Unconjugated | |
NXF1 | |
MEX67, mRNA export factor TAP, nuclear RNA export factor 1, NXF1, TAP, Tip associated protein, Tip associating protein | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10482, 53319, 59087 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation, Immunocytochemistry, Immunofluorescence | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O88984, Q99JX7, Q9UBU9 | |
Nxf1 | |
NXF1 Fusion Protein Ag0413 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 12
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
NXF1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction