missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OMA1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17116-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody is specific to OMA1.
OMA1 (Overlapping with the m-AAA protease 1 homolog) is also named as MPRP1(Metalloprotease-related protein 1) and belongs to the peptidase M48 family. It is a zinc metalloprotease that spans the inner mitochondrial membrane with a number of predicted membrane spanning domains and the 60 kDa form of OMA1 rapidly accumulated concomitantly with the cleavage of all OPA1 variants, and the 40 kDa form of OMA1 may be the inactive form, with the 60 kDa form being the active enzyme.Specifications
OMA1 | |
Polyclonal | |
Unconjugated | |
OMA1 | |
2010001O09Rik, DAB1, MPRP 1, MPRP1, OMA1, YKR087C, ZMPOMA1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
115209, 298282, 67013 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
D3ZS74, Q96E52, Q9D8H7 | |
Oma1 | |
OMA1 Fusion Protein Ag10803 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
OMA1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction