Learn More
Corning™ LSE™ Optional Head
For use with various sizes of tubes.
554.48 SEK - 1051.72 SEK
Specifications
Product Type | Optional Head |
---|---|
For Use With (Equipment) | Corning™ LSE™ Vortex Mixer |
Product Code | Brand | Holds | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Price | Quantity & Availability | ||||||
11981981
|
Corning™
480107 |
2 x 50 mL Tubes |
1051.72 SEK
Each |
Estimated Shipment: 12-11-2025
Log in to see stock. |
||||||
11961981
|
Corning™
480105 |
12 x 1.5 to 2 mL Tubes |
554.48 SEK
Each |
Estimated Shipment: 12-11-2025
Log in to see stock. |
||||||
11971981
|
Corning™
480106 |
4 x 15 mL Tubes |
1051.72 SEK
Each |
Estimated Shipment: 12-11-2025
Log in to see stock. |
||||||
Description
This is an optional head for tubes, held horizontally, for use with Corning™ LSE™ Vortex Mixer.Specifications
Optional Head | |
Corning™ LSE™ Vortex Mixer |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Corning™ LSE™ Optional Head