missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OPTN Mouse anti-Human, Clone: 6C1H4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60293-1-IG-20UL
This item is not returnable.
View return policy
Description
This gene encodes the coiled-coil containing protein optineurin. Optineurin may play a role in normal-tension glaucoma and adult-onset primary open angle glaucoma. Optineurin interacts with adenovirus E3-14.7K protein and may utilize tumor necrosis factor-alpha or Fas-ligand pathways to mediate apoptosis, inflammation or vasoconstriction. Optineurin may also function in cellular morphogenesis and membrane trafficking, vesicle trafficking, and transcription activation through its interactions with the RAB8, huntingtin, and transcription factor IIIA proteins. Alternative splicing results in multiple transcript variants encoding the same protein.Specifications
OPTN | |
Monoclonal | |
1.7 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q96CV9 | |
OPTN | |
OPTN Fusion Protein Ag1272 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunofluorescence | |
6C1H4 | |
Unconjugated | |
OPTN | |
AAT4, E3 14.7K interacting protein, FAA4, FIP 2, FIP2, GLC1E, HIP 7, HIP7, Huntingtin yeast partner L, HYPL, NEMO related protein, NRP, optineurin, OPTN | |
Mouse | |
Protein G | |
RUO | |
10133 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
OPTN Mouse anti-Human, Clone: 6C1H4, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction