missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OPTN Mouse anti-Human, Clone: 6C1H4, Proteintech
Mouse Monoclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | OPTN |
---|---|
Clone | 6C1H4 |
Concentration | 1.7 mg/mL |
Applications | Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunofluorescence |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16887365
|
Proteintech
60293-1-IG-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16877365
|
Proteintech
60293-1-IG-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene encodes the coiled-coil containing protein optineurin. Optineurin may play a role in normal-tension glaucoma and adult-onset primary open angle glaucoma. Optineurin interacts with adenovirus E3-14.7K protein and may utilize tumor necrosis factor-alpha or Fas-ligand pathways to mediate apoptosis, inflammation or vasoconstriction. Optineurin may also function in cellular morphogenesis and membrane trafficking, vesicle trafficking, and transcription activation through its interactions with the RAB8, huntingtin, and transcription factor IIIA proteins. Alternative splicing results in multiple transcript variants encoding the same protein.Specifications
OPTN | |
1.7 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
AAT4, E3 14.7K interacting protein, FAA4, FIP 2, FIP2, GLC1E, HIP 7, HIP7, Huntingtin yeast partner L, HYPL, NEMO related protein, NRP, optineurin, OPTN | |
OPTN | |
IgG1 | |
Protein G | |
Antibody |
6C1H4 | |
Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunofluorescence | |
Unconjugated | |
Mouse | |
Human | |
Q96CV9 | |
10133 | |
OPTN Fusion Protein Ag1272 | |
Primary | |
-20°C | |
OPTN |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
OPTN Mouse anti-Human, Clone: 6C1H4, Proteintech