Learn More
OPTN Mouse anti-Human, Clone: 6C1H4, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 60293-1-IG-150UL
 View more versions of this product
Product Code. 16877365
  • 4652.90 SEK / 150µL
Description
This gene encodes the coiled-coil containing protein optineurin. Optineurin may play a role in normal-tension glaucoma and adult-onset primary open angle glaucoma. Optineurin interacts with adenovirus E3-14.7K protein and may utilize tumor necrosis factor-alpha or Fas-ligand pathways to mediate apoptosis, inflammation or vasoconstriction. Optineurin may also function in cellular morphogenesis and membrane trafficking, vesicle trafficking, and transcription activation through its interactions with the RAB8, huntingtin, and transcription factor IIIA proteins. Alternative splicing results in multiple transcript variants encoding the same protein.
Specifications
Show More
Certificates
Promotions
Promotions
Product Content Correction
Product Title
OPTN Mouse anti-Human, Clone: 6C1H4, Proteintech > 150 μL; Unconjugated