Learn More
WTW™ OxiTop™-i Respirometric BOD Measuring System
The WTW™ OxiTop™-i Respirometric BOD Measuring System is designed for self-check BOD measurement with undiluted samples (according to DIN EN 1899 -2) at measuring ranges up to 4000 mg/L.
4660.00 SEK - 85085.00 SEK
Specifications
Display Type | Backlit Graphic LCD |
---|---|
Range | Measuring ranges up to 4000 mg/L BOD possible |
Product Code | Brand | Type | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Price | Quantity & Availability | ||||||
16223561
|
WTW™
208276 |
OxiTop™-i 6 |
24385.00 SEK
Each |
In Stock
Log in to see stock. |
||||||
16347231
|
WTW™
208272 |
OxiTop™-i B |
4660.00 SEK
Each |
In Stock
Log in to see stock. |
||||||
16273951
|
WTW™
208262 |
OxiTop™-i IS 12 |
63855.00 SEK
Each |
Estimated Shipment: 03-07-2025
Log in to see stock. |
||||||
16219020
|
WTW™
208260 |
OxiTop™-i IS 6 |
37045.00 SEK
Each |
In Stock
Log in to see stock. |
||||||
16308605
|
WTW™
208274 |
OxiTop™-i 2 |
8740.00 SEK
Each |
In Stock
Log in to see stock. |
||||||
16458763
|
WTW™
209700 |
OxiTop™-IDS 1 |
6230.00 SEK
Each |
In Stock
Log in to see stock. |
||||||
16391736
|
WTW™
209716 |
OxiTop™-IDS 12 |
85085.00 SEK
Pack of 12 |
In Stock
Log in to see stock. |
||||||
16367616
|
WTW™
208264 |
OxiTop™-i SET 2 |
8985.00 SEK
1 set |
Estimated Shipment: 04-07-2025
Log in to see stock. |
||||||
Description
- OxiTop™ is a respirometric vessel-top measuring system for BOD self-check measurement with undiluted samples (DIN EN 1899 -2)
- Measuring ranges up to 4000 mg/L BOD possible
- No need to convert data - direct input of sample volume
- Direct display of the BOD value on the measuring head screen
- Menu-operated, with backlit graphic LCD, control LED and operating buttons for direct BOD readings, volume pre-selection, measurement duration between 1 and 7 days selectable, display of measuring curve, display of current readings
Specifications
Backlit Graphic LCD | |
Measuring ranges up to 4000 mg/L BOD possible |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
WTW™ OxiTop™-i Respirometric BOD Measuring System