missing translation for 'onlineSavingsMsg'
Learn More
p-Phenylenediamine dihydrochloride, 99+% Product Code.: 10561724

p-Phenylenediamine dihydrochloride, 99+%

Product Code. p-183052 Shop All Thermo Scientific Chemicals Products
25 g, Glass bottle, 25g
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 10561724

Brand: Thermo Scientific Acros 417480250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 624-18-0 | C6H8N2·2HCl | 181.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 624-18-0
Molecular Formula C6H8N2·2HCl
Molecular Weight (g/mol) 181.06
MDL Number MFCD00013002
InChI Key IYXMNTLBLQNMLM-UHFFFAOYSA-N
Synonym benzene-1,4-diamine dihydrochloride, p-phenylenediamine dihydrochloride, 1,4-diaminobenzene dihydrochloride, 1,4-phenylenediamine dihydrochloride, fourrine ds, 1,4-benzenediamine, dihydrochloride, pelagol cd, pelagol grey cd, durafur black rc, fourrine 64Show MoreShow Less
PubChem CID 12205
IUPAC Name benzene-1,4-diamine;dihydrochloride
SMILES C1=CC(=CC=C1N)N.Cl.Cl

Specifications

Color Gray-Purple to White
Infrared Spectrum Authentic
Assay Percent Range 99% min. (Argentometry)
Packaging Glass bottle
Linear Formula C6H4(NH2)2·2HCl
Quantity 25 g
Solubility Information Solubility in water: soluble.
Formula Weight 181.06
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material p-Phenylenediamine dihydrochloride, 99+%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot NumberA0465879 Certificate TypeCertificate of Analysis Date11/09/2024 Product Code417481000,417480250
Lot NumberA0393596 Certificate TypeCertificate of Analysis Date14/12/2022 Product Code417481000,417480250
Lot NumberA0408510 Certificate TypeCertificate of Analysis Date13/12/2022 Product Code417481000,417480250
Lot NumberA0408511 Certificate TypeCertificate of Analysis Date13/12/2022 Product Code417481000,417480250
Lot NumberA0417053 Certificate TypeCertificate of Analysis Date12/12/2022 Product Code417481000,417480250
compliance-icons compliance-icons
Product Identifier
  • p-Phenylenediamine dihydrochloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H319-Causes serious eye irritation.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
p-Phenylenediamine dihydrochloride, 99+% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.