Learn More
P2RY1 Mouse anti-Human, Unconjugated, Clone: 1A1F3, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67654-1-IG-20UL
 View more versions of this product
Product Code. 16840406
  • 1645.54 SEK / 20µL
Description
The P2RY1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human P2RY1. This antibody recognizes human, pig, mouse, rat, rabbit antigen. The P2RY1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
P2RY1
Monoclonal
500 μg/mL
Mouse
Primary
Human, Mouse, Rat, Pig, Rabbit
Western Blot, Immunohistochemistry
1A1F3
Unconjugated
20 μL
5028
IgG1
Certificates
Promotions
Promotions
Product Content Correction
Product Title
P2RY1 Mouse anti-Human, Unconjugated, Clone: 1A1F3, Proteintech > 20 μL