Learn More
p47 phox Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 28187-1-AP-20UL
Description
The p47 phox antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human p47 phox. This antibody recognizes human antigen. The p47 phox antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
p47 phox | |
Polyclonal | |
Unconjugated | |
20 μL | |
653361 | |
IgG |
Western Blot, Immunohistochemistry | |
300 μg/mL | |
Rabbit | |
Primary | |
Human |
Product Suggestions
Customers who viewed this item also viewed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.