missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAIP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15583-1-AP-150UL
This item is not returnable.
View return policy
Specifications
PAIP2 | |
Polyclonal | |
Unconjugated | |
PAIP2 | |
PABP interacting protein 2, PAIP 2, PAIP2, PAIP2A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
361309, 51247, 67869 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q6AXZ0, Q9BPZ3, Q9D6V8 | |
Paip2 | |
PAIP2 Fusion Protein Ag7119 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PAIP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction