Learn More
Palladium AAS Standard, 1000 mg/L Pd in HCl 5%, Honeywell Fluka™
Shop All Honeywell Fluka ProductsDescription
Honeywell Fluka™ analytical standards offer a growing range of high quality inorganic and organic reference materials for the European market produced according to ISO 9001. The products are suitable for a wide range of industries and applications and come complete with a detailed Certificate of Analysis (CoA) in every delivery, and are eligible for free expert technical support ensuring you have the information you need to quickly start your analysis and pass an audit.

Specifications
Specifications
Chemical Name or Material | Palladium AAS Standard |
Density | 1.03 g/cm3 |
pH | <2 |
Quantity | 100 mL |
Grade | Standards and Modifiers for AAS |
Product Suggestions
Customers who viewed this item also viewed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.