Learn More
DiscoverX PathHunter™ eXpress GPR171 CHO-K1 β-Arrestin Orphan GPCR Assay
The PathHunter™ eXpress GPR171 CHO-K1 β-Arrestin Orphan GPCR Assay measures GPR171 (GPCR) activity via recruitment of β-Arrestin2. eXpress kits contain all assay materials: cells, reagents, and plates.
Brand:  DiscoverX 93-0415E2ACP1L
Encompass
Product Code. 17373149
  • 106850.84 SEK / 1 set
Description
The PathHunter™ GPR171 eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand based activation of GPR171 GPCR activity via detection of β-arrestin recruitment. This product uses EFC technology (www.discoverx.com/technologies-platforms/enzyme-fragment-complementation-technology). The included cells overexpress PK-tagged GPR171 and EA-tagged β-Arrestin 2. Activation of GPR171 stimulates the recruitment of β-Arrestin 2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. Orphan receptor with unknown ligand.
Specifications
Specifications
GPCR
1,000 dp (10 x 96-well)
CHO-K1
Assay-Ready eXpress Kits
GPR171
GPR171
Certificates
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
DiscoverX PathHunter™ eXpress GPR171 CHO-K1 β-Arrestin Orphan GPCR Assay