Learn More
PAX6 Mouse anti-Human, Unconjugated, Clone: 2C5A1, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67529-1-IG-20UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16800216
  • 1656.73 SEK / 20µL
Estimated Shipment: 23-04-2024
to see stock.
Description
Description
The PAX6 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PAX6. This antibody recognizes human, mouse, rat antigen. The PAX6 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
PAX6
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
2C5A1
Unconjugated
20 μL
5080
IgG1