Learn More
PAX6 Mouse anti-Human, Unconjugated, Clone: 2C5A1, Proteintech
Mouse Monoclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | PAX6 |
---|---|
Clone | 2C5A1 |
Concentration | 1000 μg/mL |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence, ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16800216
|
Proteintech
67529-1-IG-20UL |
20 μL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16890206
|
Proteintech
67529-1-IG-150UL |
150 μL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The PAX6 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PAX6. This antibody recognizes human, mouse, rat antigen. The PAX6 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
PAX6 | |
1000 μg/mL | |
Monoclonal | |
Mouse | |
5080 | |
Primary |
2C5A1 | |
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
Unconjugated | |
Human, Rat, Mouse | |
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.