Learn More
PAX6 Mouse anti-Human, Unconjugated, Clone: 2C5A1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67529-1-IG-20UL
Description
The PAX6 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PAX6. This antibody recognizes human, mouse, rat antigen. The PAX6 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
PAX6 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
2C5A1 | |
Unconjugated | |
20 μL | |
5080 | |
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.