missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PBX3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12571-1-AP-150UL
This item is not returnable.
View return policy
Description
PBX3 is a transcriptional activator that binds the sequence 5′-ATCAATCAA-3′.Specifications
PBX3 | |
Polyclonal | |
Unconjugated | |
PBX3 | |
Homeobox protein PBX3, PBX3, pre B cell leukemia homeobox 3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18516, 311876, 5090 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O35317, P40426 | |
PBX3 | |
PBX3 Fusion Protein Ag3239 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PBX3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction