missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PBX3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | PBX3 |
---|---|
Concentration | 0.13 mg/mL |
Applications | Immunofluorescence, Immunoprecipitation, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16890344
|
Proteintech
12571-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16880344
|
Proteintech
12571-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PBX3 is a transcriptional activator that binds the sequence 5′-ATCAATCAA-3′.Specifications
PBX3 | |
Immunofluorescence, Immunoprecipitation, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human, Rat, Mouse | |
O35317, P40426 | |
18516, 311876, 5090 | |
PBX3 Fusion Protein Ag3239 | |
Primary | |
-20°C | |
PBX3 |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Homeobox protein PBX3, PBX3, pre B cell leukemia homeobox 3 | |
PBX3 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PBX3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech