Learn More
PELP1 Mouse anti-Human, Unconjugated, Clone: 1B11E4, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67050-1-IG-20UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16889445
  • 1355.00 SEK / 20µL
Estimated Shipment: 31-03-2023
to see stock.
Description
Description
The PELP1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PELP1. This antibody recognizes human antigen. The PELP1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
PELP1
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Immunofluorescence, Immunohistochemistry, Western Blot
1B11E4
Unconjugated
20 μL
27043
IgG2b