Learn More
PELP1 Mouse anti-Human, Unconjugated, Clone: 1B11E4, Proteintech
Mouse Monoclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | PELP1 |
---|---|
Clone | 1B11E4 |
Concentration | 1000 ÎĽg/mL |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16889445
|
Proteintech
67050-1-IG-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16879445
|
Proteintech
67050-1-IG-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The PELP1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PELP1. This antibody recognizes human antigen. The PELP1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
PELP1 | |
1000 ÎĽg/mL | |
Monoclonal | |
Mouse | |
27043 | |
Primary |
1B11E4 | |
Western Blot, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Human | |
IgG2b |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.