Learn More
PELP1 Mouse anti-Human, Unconjugated, Clone: 1B11E4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67050-1-IG-20UL
Description
The PELP1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PELP1. This antibody recognizes human antigen. The PELP1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
PELP1 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, Immunohistochemistry, Immunofluorescence | |
1B11E4 | |
Unconjugated | |
20 μL | |
27043 | |
IgG2b |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.