missing translation for 'onlineSavingsMsg'
Learn More

PER2 Mouse anti-Human, Unconjugated, Clone: 2E6C3, Proteintech

Mouse Monoclonal Antibody

1355.00 SEK - 3665.00 SEK

Specifications

Antigen PER2
Clone 2E6C3
Concentration 1000 μg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16830186
View Documents Promotion Details
Proteintech
67513-1-IG-150UL
150 μL
3665.00 SEK
150µL
View alternative products
Save Up to 
»
Estimated Shipment: 07-12-2022
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16840186
View Documents Promotion Details
Proteintech
67513-1-IG-20UL
20 μL
1355.00 SEK
20µL
View alternative products
Save Up to 
»
Estimated Shipment: 07-12-2022
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The PER2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PER2. This antibody recognizes human, rat antigen. The PER2 antibody has been validated for the following applications: WB, ELISA analysis.

Specifications

Specifications

PER2
1000 μg/mL
Monoclonal
Mouse
8864
Primary
2E6C3
Western Blot
Unconjugated
Rat, Human
IgG1
SDS
Product Certifications