Learn More
Perforin Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14580-1-AP-20UL
 View more versions of this product
Product Code. 16883284
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
Perforin is one of the major cytolytic proteins of cytolytic granules. Perforin is a cytolytic mediator and is stored in and released by cytoplasmic granules. Moreover, perforin is involved in immune defense against tumors and virus infections as mediated by cytotoxic lymphocytes. Perforin is a 555 amino acid protein with a 21 amino acid signal peptide, and has a molecular weight of 70 to 75 kD. Perforin is a pore forming protein with a mechanism of transmembrane channel formation similar to C9, and homology between perforin and C9 have been demonstrated. Studies show that perforin is expressed only in killer cell lines and not in helper T lymphocytes or other tumor cells tested. Perforin is known to be a key effector molecule for T-cell- and natural killer-cell-mediated cytolysis. Defects in the perforin gene cause familial hemophagocytic lymphohistiocytosis type 2 (HPLH2), a rare and lethal autosomal recessive disorder of early childhood. Alternative splicing results in multiple transcript variants of perforin.
Specifications
Specifications
Perforin
Polyclonal
Unconjugated
PRF1
Cytolysin, FLH2, HPLH2, P1, Perforin, Perforin 1, PFN1, PFP, PRF1
Rabbit
Antigen Affinity Chromatography
RUO
18646, 5551
-20°C
Liquid
Western Blot
0.28 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P10820, P14222
Prf1
Perforin Fusion Protein Ag6060
20 μL
Primary
Mouse, Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Perforin Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated