missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGCP Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16601-1-AP-150UL
This item is not returnable.
View return policy
Description
PGCP is a metallopeptidase that belongs to the peptidase M28 family. The encoded protein may catalyze the cleavage of dipeptides with unsubstituted terminals into amino acids.Specifications
PGCP | |
Polyclonal | |
Unconjugated | |
CPQ | |
CPQ, LCH1, Lysosomal dipeptidase, PGCP, plasma glutamate carboxypeptidase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10404, 54381 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
0.24 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9WVJ3, Q9Y646 | |
Cpq | |
PGCP Fusion Protein Ag9770 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PGCP Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction