missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHACTR4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13408-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the phosphatase and actin regulator (PHACTR) family. Other PHACTR family members have been shown to inhibit protein phosphatase 1 (PP1) activity, and the homolog of this gene in the mouse has been shown to interact with actin and PP1. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
PHACTR4 | |
Polyclonal | |
Unconjugated | |
PHACTR4 | |
PHACTR4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
100169, 65979, 682920 | |
-20°C | |
Liquid |
Immunofluorescence, Western Blot, Immunocytochemistry | |
0.34 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q501J7, Q8IZ21 | |
PHACTR4 | |
PHACTR4 Fusion Protein Ag4054 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PHACTR4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction