Learn More
PHD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12984-1-AP-150UL
 View more versions of this product
Product Code. 16880974
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
The hypoxia inducible factor (HIF) is a transcriptional complex which is involved in oxygen homeostasis. At normal oxygen levels, the alpha subunit of HIF is targeted for degration by prolyl hydroxylation. This gene encodes an enzyme responsible for this posttranslational modification. Multiple alternatively spliced variants, encoding the same protein, have been identified.
Specifications
Specifications
PHD1
Polyclonal
Unconjugated
EGLN2
Egl nine homolog 2, EGLN2, EIT6, Estrogen induced tag 6, HIF PH1, HIF prolyl hydroxylase 1, HIFPH1, HPH 1, HPH 3, PHD1
Rabbit
Antigen Affinity Chromatography
RUO
112398, 112406, 308457
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Western Blot
0.33 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q6AYU4, Q91YE2, Q96KS0
Egln2
PHD1 Fusion Protein Ag3616
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PHD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated