missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-ZIPK (Ser311) Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 51021-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody is specific to ZIPK-Phospho 311Ser.
The protein encoded by this gene is a component of desmosomes and of the epidermal cornified envelope in keratinocytes. The N-terminal domain of this protein interacts with the plasma membrane and its C-terminus interacts with intermediate filaments. Through its rod domain, this protein forms complexes with envoplakin. This protein may serve as a link between the cornified envelope and desmosomes as well as intermediate filaments. AKT1/PKB, a protein kinase mediating a variety of cell growth and survival signaling processes, is reported to interact with this protein, suggesting a possible role for this protein as a localization signal in AKT1-mediated signaling.Specifications
Phospho-ZIPK (Ser311) | |
Polyclonal | |
Unconjugated | |
DAPK3 | |
DAP kinase 3, DAP like kinase, DAPK3, Dlk, ZIP, ZIP kinase, ZIPK | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
13144, 1613 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O43293, O54784 | |
Dapk3 | |
Peptide | |
20 ÎĽL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Phospho-ZIPK (Ser311) Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 100 ÎĽL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction