missing translation for 'onlineSavingsMsg'
Learn More

Phospho-ZIPK (Ser311) Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1287.50 SEK - 3450.50 SEK

Specifications

Antigen Phospho-ZIPK (Ser311)
Concentration 0.13 mg/mL
Applications Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16886285
View Documents
Proteintech
51021-1-AP-20UL
20 ÎĽL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16876285
View Documents
Proteintech
51021-1-AP-100UL
100 ÎĽL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This antibody is specific to ZIPK-Phospho 311Ser.

The protein encoded by this gene is a component of desmosomes and of the epidermal cornified envelope in keratinocytes. The N-terminal domain of this protein interacts with the plasma membrane and its C-terminus interacts with intermediate filaments. Through its rod domain, this protein forms complexes with envoplakin. This protein may serve as a link between the cornified envelope and desmosomes as well as intermediate filaments. AKT1/PKB, a protein kinase mediating a variety of cell growth and survival signaling processes, is reported to interact with this protein, suggesting a possible role for this protein as a localization signal in AKT1-mediated signaling.
Specifications

Specifications

Phospho-ZIPK (Ser311)
Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Human, Mouse
O43293, O54784
13144, 1613
Peptide
Primary
-20°C
DAPK3
0.13 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
DAP kinase 3, DAP like kinase, DAPK3, Dlk, ZIP, ZIP kinase, ZIPK
Dapk3
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Phospho-ZIPK (Ser311) Rabbit anti-Human, Mouse, Polyclonal, Proteintech

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.