Learn More
PHYHIPL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 16937-1-AP-150UL
 View more versions of this product
Product Code. 16826724
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
16937-1-AP detects 39 kDa and 42-45 kDa isoforms in SDS-PAGE.
PHYHIPL may play a role in the development of the central system.
Specifications
Specifications
PHYHIPL
Polyclonal
Unconjugated
PHYHIPL
KIAA1796, PHYHIPL
Rabbit
Antigen Affinity Chromatography
RUO
309901, 70911, 84457
-20°C
Liquid
Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q6AYN4, Q8BGT8, Q96FC7
Phyhipl
PHYHIPL Fusion Protein Ag10435
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PHYHIPL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated