Learn More
PLSCR4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12630-1-AP-20UL
 View more versions of this product
Product Code. 16890444
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
May mediate accelerated ATP-independent bidirectional transbilayer migration of phospholipids upon binding calcium ions that results in a loss of phospholipid asymmetry in the plasma membrane. May play a central role in the initiation of fibrin clot formation, in the activation of mast cells and in the recognition of apoptotic and injured cells by the reticuloendothelial system.
Specifications
Specifications
PLSCR4
Polyclonal
Unconjugated
PLSCR4
phospholipid scramblase 4, PL scramblase 4, PLSCR4, TRA1
Rabbit
Antigen Affinity Chromatography
RUO
235527, 300900, 57088
-20°C
Liquid
Western Blot
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P58196, Q9NRQ2
PLSCR4
PLSCR4 Fusion Protein Ag3323
20 μL
Primary
Human, Rat, Mouse
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PLSCR4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated