Learn More
Podoplanin Mouse anti-Human, Unconjugated, Clone: 1D9F3, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67432-1-IG-150UL
 View more versions of this product
Product Code. 16890056
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
The Podoplanin antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Podoplanin. This antibody recognizes human antigen. The Podoplanin antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
Specifications
Podoplanin
Monoclonal
1000 ÎĽg/mL
Mouse
Primary
Human
Immunohistochemistry, ELISA
1D9F3
Unconjugated
150 ÎĽL
10630
IgG2a
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Podoplanin Mouse anti-Human, Unconjugated, Clone: 1D9F3, Proteintech > 150 ÎĽL