missing translation for 'onlineSavingsMsg'
Learn More
Polyethylene glycol dimethacrylate, M.W. 1,000, Thermo Scientific™ Product Code.: 15432717

Polyethylene glycol dimethacrylate, M.W. 1,000, Thermo Scientific™

Product Code. 15432717
5 g, 5g
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15432717

Brand: Thermo Scientific Alfa Aesar 046534.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25852-47-5
MDL Number MFCD00081877
Synonym ethylene glycol dimethacrylate, ethylene dimethacrylate, glycol dimethacrylate, diglycol dimethacrylate, ethanediol dimethacrylate, ethylenedimethyacrylate, ethylene methacrylate, ethyldiol metacrylate, 1,2-bis methacryloyloxy ethane, ethylene glycol bis methacrylateShow MoreShow Less

Specifications

Color White
Physical Form Granular Solid
Molecular Formula CH3(CH2=)COC(OCH2CH2)nOCOC(=CH2)CH3
Solubility Information Soluble in water.
Quantity 5 g
Sensitivity Light and moisture sensitive
Chemical Name or Material Polyethylene glycol dimethacrylate, M.W. 1,000
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Polyethylene glycol dimethacrylate, M.W. 1,000, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.