Learn More
Poulten & Graf GmbH Polypropylene Adaptor for FORTUNA™ Dispenser with Thread
Complete a FORTUNA™ Dispenser with an adapter that comes in a wide selection of threads. Poulten & Graf GmbH Polypropylene Adaptor for FORTUNA™ Dispenser with Thread enables smaller containers to be supported on top the existing reservior.
149.00 SEK - 1950.00 SEK
Specifications
Autoclavable | Autoclavable |
---|---|
Color | White |
Description | Adapters for OPTIFIX and POLYFIX dispensers |
Material | Polypropylene |
For Use With (Equipment) | OPTIFIX and POLYFIX dispensers |
Product Code | Brand | Thread Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Thread Size | Price | Quantity & Availability | ||||||
10036201
|
Poulten & Graf GmbH
101 09060 |
GL 60 |
1725.00 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
10166250
|
Poulten & Graf GmbH
101 09065 |
GL 65 |
1950.00 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
10369990
|
Poulten & Graf GmbH
101 09028 |
GL 28 |
149.00 SEK
Each |
Estimated Shipment: 14-07-2025
Log in to see stock. |
||||||
10024750
|
Poulten & Graf GmbH
101 09030 |
GL 30 |
149.00 SEK
Each |
Estimated Shipment: 14-07-2025
Log in to see stock. |
||||||
10496881
|
Poulten & Graf GmbH
101 09036 |
GL 36 |
231.00 SEK
Each |
Estimated Shipment: 14-07-2025
Log in to see stock. |
||||||
10390131
|
Poulten & Graf GmbH
101 09033 |
GL 33 |
231.00 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
10646171
|
Poulten & Graf GmbH
101 09055 |
GL 55 |
1650.00 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
10610713
|
Poulten & Graf GmbH
101 09025 |
GL 25 |
149.00 SEK
Each |
Estimated Shipment: 29-07-2025
Log in to see stock. |
||||||
Description
- Suitable for OPTIFIX and POLYFIX dispenser
- Made of Polypropylene
- Adapter with threads
- Can be used for most chemicals and liquids
Specifications
Autoclavable | |
Adapters for OPTIFIX and POLYFIX dispensers | |
OPTIFIX and POLYFIX dispensers |
White | |
Polypropylene | |
1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Poulten & Graf GmbH Polypropylene Adaptor for FORTUNA™ Dispenser with Thread