Learn More
BRAND™ Impact-Resistant Polystyrene Laboratory Spatulas
Constructed of impact-resistant polystyrene. Available with double spatulate ends or a spatulate and spoon combination.
Brand: BRAND™ 139810
300.74 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Impact-resistant polystyrene construction
- Green in color
- Supplied in packs of 10
Specifications
10 Pk. | |
Green | |
Polystyrene |
Double spatula | |
Spatula | |
150 mm |
Product Content Correction
Product Title
BRAND™ Impact-Resistant Polystyrene Laboratory Spatulas > Length: 150mm
Spot an opportunity for improvement?Share a Content Correction