missing translation for 'onlineSavingsMsg'
Learn More

POTEH Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 24837-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16883335

  • 1355.00 SEK / 20µL
Estimated Shipment: 03-04-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

POTEH belongs to the POTE family and contains 7 ANK repeats.
Specifications

Specifications

POTEH
Polyclonal
Unconjugated
POTEH
A26C3, ACTBL1, ANKRD26 like family C member 3, POTE 22, POTE22, POTEH
Rabbit
Antigen Affinity Chromatography
RUO
23784
-20°C
Liquid
Western Blot
0.28 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q6S545
POTEH
POTEH Fusion Protein Ag20275
20 μL
Primary
Human
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers