Learn More
3M™ Powered Air Respirator System Starter Kit PF-619E
PAPR Starter Kit PF-619E is a durable powered air respirator system designed for heavy duty environments at an affordable price. This starter kit includes a turbo unit, battery, charger and belt.
Brand: 3M™ 7100236675
This item is not returnable.
View return policy
Description
- Starter Kit includes a 3M™ PF-600E turbo unit, charger and belt.
- Compatible with the 3M™ DT Series filters and specific 3M™ Versaflo™ headtops
- Audible and visual alarms alert the user to low battery or if the flow rate falls below the manufacturer's minimum design flow rate
- Durable design suitable for cleaning by shower/water spray
- Integrated battery and simple one-button functionality
Specifications
4 to 8 hours | |
3M™ Powered Air Respirator System Starter Kit PF-619E | |
No Headgear Included | |
PF-619E |
Lithium-ion | |
PAPR Assembly, No Headtop | |
No |
Product Content Correction
Product Title
3M™ Powered Air Respirator System Starter Kit PF-619E > PAPR System; Design Type: Belt-Mounted
Spot an opportunity for improvement?Share a Content Correction