missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPAPDC2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26234-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
PPAPDC2 is a phosphatase that dephosphorylates Presqualene diphosphate (PSDP) into presqualene monophosphate (PSMP), suggesting that it may have important role in innate immunity. PSDP is a bioactive lipid that rapidly remodels to PSMP upon cell activation. PPAPDC2 displays diphosphate phosphatase activity with a substrate preference for PSDP > FDP > phosphatidic acid. PPAPDC2 activity is independent of Mg2+ and has been identified in activated human neutrophil (PMN) extracts. It is widely expressed in human tissues. Recent studies shows PPAPDC2 is a functional isoprenoid diphosphate phosphatase.Specifications
PPAPDC2 | |
Polyclonal | |
Unconjugated | |
PLPP6 | |
PPAPDC2, PSDP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
403313 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunoprecipitation, Immunofluorescence | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8IY26 | |
PLPP6 | |
PPAPDC2 Fusion Protein Ag23856 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |