Learn More
PPAR Gamma Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22061-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
The PPAR Gamma antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human PPAR Gamma. This antibody recognizes human, mouse, rat antigen. The PPAR Gamma antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
PPAR Gamma | |
Polyclonal | |
Unconjugated | |
20 μL | |
5468 | |
IgG |
ELISA, Immunohistochemistry, Western Blot | |
407 μg/mL | |
Rabbit | |
Primary | |
Mouse, Rat, Human |