Learn More
PPAR Gamma Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1706.43 SEK - 4863.32 SEK
Specifications
Antigen | PPAR Gamma |
---|---|
Concentration | 407 ÎĽg/mL |
Applications | Western Blot, Immunohistochemistry, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
The PPAR Gamma antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human PPAR Gamma. This antibody recognizes human, mouse, rat antigen. The PPAR Gamma antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
PPAR Gamma | |
Western Blot, Immunohistochemistry, ELISA | |
Unconjugated | |
Human, Rat, Mouse | |
IgG |
407 ÎĽg/mL | |
Polyclonal | |
Rabbit | |
5468 | |
Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.