Learn More
PPAR Gamma Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 22061-1-AP-20UL
 View more versions of this product
Product Code. 16881605
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
This item is not returnable. View return policy
Description
Description
The PPAR Gamma antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human PPAR Gamma. This antibody recognizes human, mouse, rat antigen. The PPAR Gamma antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
PPAR Gamma
Polyclonal
Unconjugated
20 μL
5468
IgG
Western Blot, Immunohistochemistry, ELISA
407 μg/mL
Rabbit
Primary
Human, Rat, Mouse
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PPAR Gamma Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech > 20 μL