missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRAP1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11932-1-AP-20UL
This item is not returnable.
View return policy
Description
Proline rich acidic protein 1 (PRAP1) is also named as PRO1195 or UPA. It may play an important role in maintaining normal growth homeostasis in epithelial cells. Mitotic arrest deficient 1 (MAD1) is a key component in mitotic checkpoint signaling. Recent study identified PRAP1 as a novel MAD1 binding partner using yeast-two hybrid screening. PRAP1 has four isoforms with MW 16-20 kDa and 10 kDa.Specifications
PRAP1 | |
Polyclonal | |
Unconjugated | |
PRAP1 | |
PRAP1, PRO1195, proline rich acidic protein 1, RP11 122K13.6, UPA | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
118471, 22264 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.34 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q80XD8, Q96NZ9 | |
PRAP1 | |
PRAP1 Fusion Protein Ag2526 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PRAP1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction