missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRPF40B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16929-1-AP-150UL
This item is not returnable.
View return policy
Specifications
PRPF40B | |
Polyclonal | |
Unconjugated | |
PRPF40B | |
Huntingtin yeast partner C, HYPC, PRPF40B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
25766, 363000, 54614 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6NWY9, Q80W14 | |
Prpf40b | |
PRPF40B Fusion Protein Ag10361 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PRPF40B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction