Learn More
PSMC2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14905-1-AP-20UL
 View more versions of this product
Product Code. 16883804
Description
Description
May be involved in transcriptional regulation.
Specifications
Specifications
PSMC2
Polyclonal
Unconjugated
PSMC2
MSS1, Nbla10058, Protein MSS1, PSMC2, S7
Rabbit
Antigen Affinity Chromatography
RUO
19181, 25581, 5701
-20°C
Liquid
Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunofluorescence
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P35998, P46471, Q63347
Psmc2
PSMC2 Fusion Protein Ag6700
20 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
PSMC2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated