Learn More
PTBP1 Mouse anti-Human, Unconjugated, Clone: 3D6D8, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67462-1-IG-20UL
 View more versions of this product
Product Code. 16800106
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
The PTBP1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PTBP1. This antibody recognizes human, mouse, rat antigen. The PTBP1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
PTBP1
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
3D6D8
Unconjugated
20 μL
5725
IgG1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PTBP1 Mouse anti-Human, Unconjugated, Clone: 3D6D8, Proteintech > 20 μL