missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTH Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19001-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
PTH is a hormone producted by the parathyroid gland that regulates the concentration of calcium and phosphorus in extracellular fluid. It is produced in the parathyroid gland as an 84 amino acid single chain polypeptide. Defects in this gene are a cause of familial isolated hypoparathyroidism.Specifications
PTH | |
Polyclonal | |
Unconjugated | |
PTH | |
Parathormone, Parathyrin, parathyroid hormone, PTH, PTH1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
5741 | |
-20°C | |
Liquid |
Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P01270 | |
PTH | |
Peptide | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |